Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145960.1 | NP_001139432.1 | 17 | 1822 | ||
NM_020258.4 | NP_064654.3 | 17 | 1822 | |||
GenBank mRNA | AF121081.1 | - | 17 | 1721 | ||
AK028984.1 | - | 17 | 1498 | |||
AK030881.1 | - | 5 | 1029 | |||
AK031045.1 | - | 17 | 1760 | |||
AK079414.1 | - | 17 | 1498 | |||
AK079823.1 | - | 17 | 1823 | |||
AK220151.1 | - | 15 | 1264 | |||
BC022752.1 | - | 3 | 197 | |||
BC038000.1 | - | 17 | 1493 | |||
BC048836.1 | - | 3 | 137 | |||
BC063326.1 | - | 17 | 1510 | 30301217 |