Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020295.3 | NP_064691.2 | 3, 4 | 385 | ||
GenBank mRNA | AF190665.1 | - | 3, 4 | 187 | ||
AK030631.1 | - | 4, 5 | 1191 | |||
AK178182.1 | - | 3, 4 | 214 | |||
BC016110.1 | - | 3, 4 | 225 | 4481088 |