Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020295.3 | NP_064691.2 | 10 | 980 | ||
GenBank mRNA | AF190665.1 | - | 10 | 782 | ||
AK030631.1 | - | 11 | 1786 | |||
AK195974.1 | - | 3 | 171 | |||
BC016110.1 | - | 10 | 739 | 4481088 |