Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039386.1 | NP_001034475.1 | 12 | 1410 | ||
NM_001039387.1 | NP_001034476.1 | 11 | 1404 | |||
NM_001177654.1 | NP_001171125.1 | 11 | 1404 | |||
NM_001177655.1 | NP_001171126.1 | 10 | 1314 | |||
NM_020276.3 | NP_064672.2 | 11 | 1341 | |||
GenBank mRNA | AF266508.2 | - | 10 | 1307 | ||
AJ278902.1 | - | 12 | 1188 | |||
AK004986.2 | - | 11 | 1401 | |||
AK019560.1 | - | 12 | 1377 | |||
AK053402.1 | - | 10 | 1261 | |||
AK158803.1 | - | 12 | 1140 | |||
AK165300.1 | - | 11 | 1349 | |||
BC006642.1 | - | 11 | 1362 | 3483536 |