Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 5 | 672 | ||
NM_001141932.1 | NP_001135404.1 | 5 | 672 | |||
NM_020296.2 | NP_064692.2 | 5 | 865 | |||
GenBank mRNA | AB026569.1 | - | 5 | 588 | ||
AK133054.1 | - | 5 | 393 | |||
AK135280.1 | - | 5 | 552 | |||
AK145897.1 | - | 5 | 674 | |||
AK147109.1 | - | 5 | 866 | |||
AK161102.1 | - | 5 | 622 | |||
AK164307.1 | - | 5 | 666 | |||
BC016501.1 | - | 5 | 853 | 4527742 | ||
BC056609.1 | - | 2 | 64 | |||
BC057866.1 | - | 5 | 704 | 5327143 |