Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141931.1 | NP_001135403.1 | 8 | 1027 | ||
NM_001141932.1 | NP_001135404.1 | 8 | 1027 | |||
NM_020296.2 | NP_064692.2 | 8 | 1220 | |||
GenBank mRNA | AB026569.1 | - | 8 | 943 | ||
AK133054.1 | - | 8 | 748 | |||
AK135280.1 | - | 8 | 907 | |||
AK145897.1 | - | 8 | 1029 | |||
AK147109.1 | - | 8 | 1221 | |||
AK161102.1 | - | 8 | 977 | |||
AK161416.1 | - | 4 | 357 | |||
AK164307.1 | - | 8 | 1021 | |||
AK178547.1 | - | 2 | 58 | |||
AK180025.1 | - | 1 | 59 | |||
AK188295.1 | - | 2 | 58 | |||
AK210454.1 | - | 2 | 87 | |||
BC016501.1 | - | 8 | 1208 | 4527742 | ||
BC056609.1 | - | 5 | 419 | |||
BC057866.1 | - | 8 | 1059 | 5327143 |