Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252617.1 | NP_001239546.1 | 2 | 162 | ||
NM_001252618.1 | NP_001239547.1 | 2 | 162 | |||
NM_001252619.1 | NP_001239548.1 | 2 | 162 | |||
NM_002297.3 | NP_002288.1 | 2 | 162 | |||
XM_011518675.2 | XP_011516977.1 | 2 | 304 | |||
GenBank mRNA | BC065721.1 | - | 2 | 164 | 30330221 | |
BC074925.2 | - | 2 | 126 | 30915261 | ||
BC074926.2 | - | 2 | 126 | 30915486 | ||
CD721968.1 | - | 2 | 142 | |||
CK429694.1 | - | 2 | 139 | |||
CK430667.1 | - | 2 | 150 | |||
X62418.1 | - | 2 | 124 | |||
X67647.1 | - | 2 | 145 |