Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dolpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290508.1 | NP_001277437.1 | 1 | 241 | ||
NM_001290509.1 | NP_001277438.1 | 1 | 241 | |||
NM_020329.4 | NP_065062.1 | 1 | 241 | |||
GenBank mRNA | AB030189.1 | - | 1 | 81 | ||
AK002544.1 | - | 1 | 80 | |||
AK044141.1 | - | 1 | 80 | |||
AK053923.1 | - | 1 | 70 | |||
AK088065.1 | - | 1 | 85 | |||
BC026544.1 | - | 1 | 65 | 4975127 | ||
BY133699.1 | - | 1 | 166 |