Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020498.2 | NP_065244.2 | 3 | 655 | ||
GenBank mRNA | AF232024.1 | - | 3 | 577 | ||
BC125350.1 | - | 3 | 655 | 40129865 | ||
BC132052.1 | - | 3 | 655 | 40130329 | ||
BC145087.1 | - | 4 | 397 | 9053635 |