Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020503.2 | NP_065249.2 | 1 | 873 | ||
GenBank mRNA | BC111811.1 | - | 1 | 874 | ||
BC128078.1 | - | 1 | 870 | |||
BC128079.1 | - | 1 | 874 | |||
BC152359.1 | - | 1 | 874 | 40101839 |