Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020509.3 | NP_065255.2 | 2, 3 | 234 | ||
GenBank mRNA | AF205951.1 | - | 1, 2 | 167 | ||
AF290872.1 | - | 2, 3 | 272 | |||
AF316397.2 | - | 2, 3 | 176 | |||
AF323082.1 | - | 2, 3 | 244 | |||
AK007553.1 | - | 2, 3 | 179 | |||
BC029248.1 | - | 1, 2 | 174 | 4189285 |