Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019568.2 | NP_062514.2 | 4 | 741 | ||
GenBank mRNA | AB041614.1 | - | 4 | 741 | ||
AF144754.1 | - | 4 | 565 | |||
AF152377.1 | - | 4 | 491 | |||
AF192557.1 | - | 4 | 739 | |||
AF252873.1 | - | 4 | 739 | |||
AF352785.1 | - | 4 | 351 | |||
AK004615.1 | - | 4 | 585 | |||
AK014351.1 | - | 4 | 531 | |||
AK076112.1 | - | 4 | 583 | |||
AK155356.1 | - | 4 | 579 | |||
AU035952.1 | - | 4 | 738 | |||
BC079661.1 | - | 3 | 381 | |||
BC147274.1 | - | 4 | 485 | 9056321 | ||
BC147275.1 | - | 4 | 485 | 9056329 |