Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005565.3 | NP_005556.1 | 3, 4 | 401 | ||
GenBank mRNA | AK292890.1 | - | 3, 4 | 362 | ||
AK300000.1 | - | 3, 4 | 380 | |||
AK310369.1 | - | 3, 4 | 342 | |||
BC016618.1 | - | 3, 4 | 338 | 4476274 | ||
BT007273.1 | - | 3, 4 | 194 | |||
FN432830.1 | - | 3, 4 | 194 | |||
U20158.1 | - | 3, 4 | 449 |