Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026545.3 | NP_080821.3 | 4 | 639 | ||
GenBank mRNA | AK002921.1 | - | 4 | 478 | ||
AK003436.1 | - | 4 | 480 | |||
AK020132.1 | - | 4 | 498 | |||
AK027973.1 | - | 4 | 443 | |||
AK051257.1 | - | 4 | 491 | |||
AK150493.1 | - | 4 | 482 | |||
AK150982.1 | - | 4 | 446 | |||
AK153329.1 | - | 4 | 480 | |||
AK159216.1 | - | 4 | 448 | |||
AK159795.1 | - | 4 | 464 | |||
AK159926.1 | - | 4 | 474 | |||
AK166718.1 | - | 4 | 473 | |||
AK167948.1 | - | 4 | 448 | |||
AK168886.1 | - | 4 | 447 | |||
BC004075.1 | - | 4 | 457 | 3591796 | ||
BC005717.1 | - | 4 | 367 | 2609781 | ||
CF732286.1 | - | 4 | 639 |