Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gigyf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031408.2 | NP_113596.2 | 17 | 2057 | ||
GenBank mRNA | AK220289.1 | - | 10 | 1066 | ||
AY176042.1 | - | 17 | 2073 | |||
BC052875.1 | - | 11 | 1161 |