Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174097.2 | NP_001167568.1 | 4 | 725 | ||
NM_001315537.1 | NP_001302466.1 | 4 | 503 | |||
NM_002300.7 | NP_002291.1 | 4 | 503 | |||
XM_006719074.2 | XP_006719137.1 | 4 | 619 | |||
GenBank mRNA | AK129881.1 | - | 4 | 454 | ||
AK312328.1 | - | 4 | 487 | |||
AL516910.3 | - | 4 | 724 | |||
AY189689.1 | - | 4 | 479 | |||
BC002362.1 | - | 4 | 470 | 2961445 | ||
BC015122.1 | - | 4 | 494 | 3685985 | ||
BC071860.1 | - | 4 | 435 | 4902461 | ||
BT019764.1 | - | 3 | 392 | |||
BT019765.1 | - | 3 | 392 | |||
BU540074.1 | - | 4 | 506 | |||
CX867875.1 | - | 4 | 700 | |||
EU794633.1 | - | 4 | 446 | |||
Y00711.1 | - | 4 | 446 |