Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020579.2 | NP_065604.2 | 3, 4 | 553 | ||
GenBank mRNA | AF142671.1 | - | 3, 4 | 493 | ||
AK155471.1 | - | 4, 5 | 653 | |||
BC004095.1 | - | 3, 4 | 312 | |||
BC013619.1 | - | 3, 4 | 526 | 3965561 | ||
BU703242.1 | - | 3, 4 | 552 |