Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020619.2 | NP_065644.2 | 4 | 2294 | ||
GenBank mRNA | AK007528.1 | - | 1 | 498 | ||
AK150873.1 | - | 1 | 1056 | |||
AK151941.1 | - | 1 | 1030 | |||
AK152127.1 | - | 1 | 1030 | |||
AK157286.1 | - | 4 | 2291 | |||
AK195586.1 | - | 1 | 87 | |||
AK217769.1 | - | 1 | 87 | |||
AK218085.1 | - | 1 | 87 | |||
BC051949.1 | - | 4 | 2294 | 5716189 |