Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002301.4 | NP_002292.1 | 6 | 864 | ||
NM_017448.3 | NP_059144.1 | 6 | 810 | |||
XM_017017721.1 | XP_016873210.1 | 5 | 754 | |||
XM_017017722.1 | XP_016873211.1 | 5 | 675 | |||
GenBank mRNA | AF401094.1 | - | 5 | 638 | ||
AF401095.1 | - | 4 | 464 | |||
AF401097.1 | - | 6 | 756 | |||
AK058181.1 | - | 6 | 786 | |||
AY286300.1 | - | 6 | 786 | |||
BC019249.2 | - | 6 | 817 | 4839028 | ||
BC064388.1 | - | 6 | 833 | 5744600 | ||
BC090043.1 | - | 6 | 762 | 30387780 | ||
GU479374.1 | - | 5 | 702 | |||
HQ386000.1 | - | 6 | 738 | |||
HQ386001.1 | - | 6 | 792 | |||
HQ386002.1 | - | 5 | 674 | |||
HQ386004.1 | - | 5 | 620 | |||
J02938.1 | - | 6 | 764 | |||
U13680.1 | - | 6 | 839 |