Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020594.2 | NP_065619.2 | 7 | 888 | ||
GenBank mRNA | AF061961.1 | - | 7 | 896 | ||
AK132679.1 | - | 6 | 1670 | |||
AK164491.1 | - | 6 | 1670 | |||
BC048687.1 | - | 7 | 820 | 6334861 |