Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarapl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020590.4 | NP_065615.1 | 4 | 563 | ||
GenBank mRNA | AB041648.1 | - | 4 | 587 | ||
AF180518.1 | - | 4 | 590 | |||
AK006103.1 | - | 4 | 362 | |||
AK011975.1 | - | 4 | 558 | |||
AK014083.1 | - | 4 | 377 | |||
AK015049.1 | - | 4 | 380 | |||
AK018687.1 | - | 4 | 564 | |||
AK159015.1 | - | 4 | 558 | |||
AK167500.1 | - | 4 | 556 | |||
AK168921.1 | - | 4 | 595 | |||
BC004602.1 | - | 4 | 498 | 3585132 | ||
BC024706.1 | - | 4 | 562 | 3711973 |