Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042484.1 | NP_001035949.1 | 3 | 229 | ||
NM_020585.2 | NP_065610.2 | 2 | 442 | |||
GenBank mRNA | AB041568.1 | - | 2 | 376 | ||
AK048175.1 | - | 3 | 205 | |||
AK077582.1 | - | 3 | 203 | |||
AK089856.1 | - | 3 | 217 | |||
AK150462.1 | - | 3 | 223 | |||
AK150696.1 | - | 2 | 384 | |||
AK151353.1 | - | 3 | 223 | |||
AK152520.1 | - | 3 | 223 | |||
AK152606.1 | - | 3 | 202 | |||
AK153213.1 | - | 3 | 223 | |||
AK158227.1 | - | 2 | 348 | |||
AK170129.1 | - | 3 | 208 | |||
AK179247.1 | - | 2 | 52 | |||
AK184095.1 | - | 2 | 51 | |||
BC016894.1 | - | 3 | 181 | 4223937 | ||
BC019697.1 | - | 2 | 367 | |||
BC058523.1 | - | 2 | 286 | 5717934 | ||
BC093518.1 | - | 3 | 199 | 3157520 | ||
BC103796.1 | - | 3 | 194 | 6519018 | ||
BY330336.1 | - | 3 | 231 |