Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem183a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042485.1 | NP_001035950.1 | 7 | 937 | ||
NM_020588.2 | NP_065613.1 | 7 | 940 | |||
GenBank mRNA | AB041592.1 | - | 7 | 896 | ||
AK004922.2 | - | 7 | 921 | |||
AK007779.1 | - | 7 | 919 | |||
AK152852.1 | - | 7 | 880 | |||
AK168331.1 | - | 7 | 882 | |||
BC003210.1 | - | 7 | 865 | 3156957 |