Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000230.2 | NP_000221.1 | 3 | 312 | ||
XM_005250340.4 | XP_005250397.1 | 3 | 386 | |||
GenBank mRNA | AF008123.1 | - | 2 | 272 | ||
AK313553.1 | - | 3 | 310 | |||
BC060830.1 | - | 3 | 310 | 30333193 | ||
BC069323.1 | - | 2 | 306 | 7262097 | ||
BC069452.1 | - | 2 | 306 | 7262109 | ||
BC069527.1 | - | 2 | 306 | 7262121 | ||
D49487.1 | - | 2 | 269 | |||
U18915.1 | - | 2 | 255 | |||
U43653.1 | - | 2 | 311 |