Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021310.3 | NP_067285.2 | 4 | 1853 | ||
GenBank mRNA | AB221690.1 | - | 4 | 1351 | ||
AF201390.1 | - | 4 | 1477 | |||
AK139934.1 | - | 4 | 1656 | |||
AK141957.1 | - | 4 | 1854 | |||
BC069906.1 | - | 4 | 1707 | |||
BC090835.1 | - | 4 | 1734 | 5705427 |