Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021311.3 | NP_067286.1 | 19 | 2288 | ||
GenBank mRNA | AB032604.1 | - | 19 | 2253 | ||
AF438405.1 | - | 19 | 2288 | |||
BC129857.1 | - | 18 | 2138 | 40109791 | ||
BC129858.1 | - | 18 | 2138 | 40109793 | ||
EF196092.1 | - | 17 | 2098 |