Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021315.2 | NP_067290.2 | 10 | 1372 | ||
GenBank mRNA | AB077991.1 | - | 10 | 1335 | ||
AK089396.1 | - | 1 | 2254 | |||
AK134051.1 | - | 10 | 1330 | |||
AK168944.1 | - | 10 | 1373 | |||
BC031132.1 | - | 10 | 1335 | 4954951 |