Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271476.1 | NP_001258405.1 | 3 | 1134 | ||
NM_001271477.1 | NP_001258406.1 | 2 | 1026 | |||
NM_001271478.1 | NP_001258407.1 | 3 | 1134 | |||
NM_001271479.1 | NP_001258408.1 | 2 | 1026 | |||
NM_021319.5 | NP_067294.2 | 2 | 1026 | |||
GenBank mRNA | AB084140.1 | - | 3 | 1134 | ||
AB084141.1 | - | 3 | 1134 | |||
AF149837.1 | - | 2 | 1026 | |||
AF149838.1 | - | 2 | 1026 | |||
AF315578.1 | - | 2 | 1026 | |||
AF315584.1 | - | 2 | 1026 | |||
AF317548.1 | - | 2 | 1026 | |||
AF392055.1 | - | 3 | 1060 | |||
AK039953.1 | - | 3 | 1134 | |||
AK050019.1 | - | 2 | 1027 | |||
AY282722.1 | - | 2 | 964 | |||
BC019396.1 | - | 2 | 999 | 5124925 | ||
EF204951.1 | - | 2 | 914 |