Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB162929.1 | - | 4 | 631 | ||
AF190699.1 | - | 4 | 703 | |||
AF190991.1 | - | 4 | 703 | |||
AK015486.1 | - | 3 | 354 | |||
AK018148.1 | - | 4 | 695 | |||
AK031550.1 | - | 4 | 731 | |||
AK042758.1 | - | 4 | 484 | |||
AK043998.1 | - | 4 | 751 | |||
AK048413.1 | - | 4 | 702 | |||
AK049903.1 | - | 3 | 437 | |||
AK081981.1 | - | 4 | 704 | |||
AK083126.1 | - | 3 | 828 | |||
BC046310.1 | - | 4 | 689 | 6307420 | ||
BC065694.1 | - | 4 | 695 | 6813365 |