Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077698.1 | NP_001071166.1 | 17 | 2194 | ||
NM_019679.2 | NP_062653.2 | 17 | 2194 | |||
GenBank mRNA | AF006466.1 | - | 17 | 1941 | ||
AF215666.1 | - | 17 | 2019 | |||
AK131173.1 | - | 11 | 1330 | |||
AK158820.1 | - | 5 | 1157 |