Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199275.2 | NP_001186204.1 | 15 | 1774 | ||
NM_001199276.2 | NP_001186205.1 | 13 | 1444 | |||
NM_001271455.1 | NP_001258384.1 | 14 | 1497 | |||
NM_001271456.1 | NP_001258385.1 | 15 | 1669 | |||
NM_021327.4 | NP_067302.2 | 14 | 1616 | |||
GenBank mRNA | AJ242777.1 | - | 13 | 1416 | ||
AJ242778.1 | - | 14 | 1588 | |||
AK135338.1 | - | 13 | 1379 | |||
AK156131.1 | - | 14 | 1470 | |||
AK170841.1 | - | 13 | 1422 | |||
AK172890.1 | - | 1402 | ||||
AK188071.1 | - | 3 | 220 | |||
AK194753.1 | - | 3 | 189 | |||
AK195559.1 | - | 4 | 341 | |||
AK217584.1 | - | 1 | 50 | |||
AY012159.2 | - | 11 | 1186 | |||
BC008186.1 | - | 14 | 1533 | 3589311 | ||
BC008665.1 | - | 15 | 1640 | 3598934 | ||
CB599739.1 | - | 5 | 503 |