Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040167.1 | NP_001035257.1 | 1 | 376 | ||
NM_001040168.1 | NP_001035258.1 | 1 | 376 | |||
GenBank mRNA | AF193612.1 | - | 1 | 51 | ||
U94354.1 | - | 1 | 49 |