Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301692.1 | NP_001288621.1 | 4 | 554 | ||
NM_001301694.1 | NP_001288623.1 | 4 | 504 | |||
NM_021340.4 | NP_067315.1 | 5 | 676 | |||
GenBank mRNA | AF076930.1 | - | 5 | 596 | ||
AK044450.1 | - | 5 | 628 | |||
AK044649.1 | - | 5 | 627 | |||
AK149272.1 | - | 5 | 627 | |||
BC082611.1 | - | 5 | 613 | |||
BC086689.1 | - | 5 | 603 | |||
BC119055.1 | - | 5 | 610 | 8733804 | ||
BC144937.1 | - | 4 | 489 | |||
CK623457.1 | - | 4 | 435 | |||
CK626179.1 | - | 4 | 492 |
Set Membership: |
G-protein coupled receptor |