Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021342.1 | NP_067317.1 | 2 | 179 | ||
GenBank mRNA | AF076533.1 | - | 2 | 179 | ||
AK133469.1 | - | 2 | 218 | |||
BC032920.1 | - | 2 | 207 | 3472318 | ||
BC034849.1 | - | 2 | 201 |