Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020581.2 | NP_065606.2 | 2 | 585 | ||
GenBank mRNA | AB054540.1 | - | 2 | 471 | ||
AF123261.1 | - | 2 | 516 | |||
AF169313.1 | - | 2 | 448 | |||
AF278699.1 | - | 2 | 520 | |||
AK009161.1 | - | 2 | 291 | |||
AK014564.1 | - | 586 | ||||
AK032491.1 | - | 2 | 554 | |||
AK132761.1 | - | 2 | 554 | |||
AK134419.1 | - | 2 | 554 | |||
BC006611.1 | - | 2 | 537 | 3966729 | ||
BC021343.1 | - | 2 | 551 | 4194173 | ||
BC025797.1 | - | 2 | 521 | 5137159 | ||
KJ650202.1 | - | 2 | 402 |