Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034029.1 | NP_001029201.1 | 8, 9 | 886 | ||
NM_001034031.1 | NP_001029203.1 | 10, 11 | 961 | |||
NM_145826.5 | NP_665825.2 | 9, 10 | 1313 | |||
GenBank mRNA | AF458068.1 | - | 9, 10 | 963 | ||
AK046302.1 | - | 9, 10 | 1316 | |||
AK046713.1 | - | 9, 10 | 975 | |||
AK078876.1 | - | 2, 3 | 1129 | |||
AK169498.1 | - | 9, 10 | 1141 | |||
BC069861.1 | - | 9, 10 | 990 | 6311568 | ||
DQ092339.1 | - | 9, 10 | 902 | |||
DQ092340.1 | - | 8, 9 | 886 | |||
DQ092341.1 | - | 10, 11 | 961 |