Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034029.1 | NP_001029201.1 | 14 | 1855 | ||
NM_001034031.1 | NP_001029203.1 | 16 | 1930 | |||
NM_145826.5 | NP_665825.2 | 15 | 2282 | |||
GenBank mRNA | AF458068.1 | - | 14 | 1872 | ||
AK046302.1 | - | 15 | 2285 | |||
AK046713.1 | - | 15 | 1944 | |||
AK078876.1 | - | 7 | 2198 | |||
AK169498.1 | - | 15 | 2110 | |||
BC026737.1 | - | 5 | 1026 | |||
BC069861.1 | - | 14 | 1821 | 6311568 | ||
DQ092339.1 | - | 15 | 1871 | |||
DQ092340.1 | - | 14 | 1855 | |||
DQ092341.1 | - | 16 | 1930 |