Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sertad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038625.1 | NP_001033714.1 | 2 | 488 | ||
NM_021372.2 | NP_067347.2 | 2 | 699 | |||
GenBank mRNA | AB041541.1 | - | 2 | 618 | ||
AK076787.1 | - | 2 | 392 | |||
AK132846.1 | - | 2 | 468 | |||
AK132849.1 | - | 2 | 307 | |||
AK142018.1 | - | 2 | 468 | |||
AK146621.1 | - | 2 | 489 | |||
AK146897.1 | - | 2 | 489 | |||
AK147674.1 | - | 3 | 875 | |||
AK184872.1 | - | 2 | 235 | |||
BC014726.1 | - | 2 | 531 | 4240282 |