Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167730.1 | NP_001161202.1 | 5 | 363 | ||
NM_021385.2 | NP_067360.2 | 5 | 363 | |||
GenBank mRNA | AF205278.1 | - | 5 | 287 | ||
AK012795.1 | - | 5 | 364 | |||
AK030043.1 | - | 5 | 364 | |||
AK048975.1 | - | 5 | 338 | |||
BC011120.1 | - | 5 | 326 | 4014013 |