Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135727.2 | NP_001129199.1 | 6 | 1107 | ||
NM_021389.6 | NP_067364.2 | 5 | 672 | |||
GenBank mRNA | AK004636.1 | - | 5 | 672 | ||
AK018032.1 | - | 5 | 688 | |||
AK020782.1 | - | 5 | 649 | |||
AK030839.1 | - | 5 | 673 | |||
AK038540.1 | - | 8 | 971 | |||
AK049182.1 | - | 6 | 1108 | |||
AK080930.1 | - | 6 | 972 | |||
AK172022.1 | - | 6 | 1088 | |||
BC034646.1 | - | 5 | 676 | |||
BC052863.1 | - | 5 | 621 |