Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021393.2 | NP_067368.2 | 8 | 1303 | ||
GenBank mRNA | AB041607.1 | - | 7 | 1424 | ||
AK080419.1 | - | 8 | 1005 | |||
AK087524.1 | - | 8 | 1003 | |||
AK173124.1 | - | 7 | 838 | |||
AK179149.1 | - | 3 | 178 | |||
AK189417.1 | - | 3 | 177 | |||
AK214584.1 | - | 3 | 177 | |||
BC004762.1 | - | 8 | 1191 | 3155904 |