Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026907.3 | NP_081183.3 | 5 | 762 | ||
GenBank mRNA | AF210700.1 | - | 4 | 665 | ||
AK007321.1 | - | 4 | 624 | |||
AK029082.1 | - | 5 | 763 | |||
AK143807.1 | - | 5 | 748 | |||
BC010805.1 | - | 5 | 839 | 4239879 | ||
BC020159.1 | - | 4 | 710 | 4016267 |