Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159697.1 | NP_001153169.1 | 1 | 585 | ||
GenBank mRNA | AB041550.1 | - | 1 | 585 | ||
BC139438.1 | - | 1 | 582 | 8862460 | ||
BC139439.1 | - | 1 | 581 | 8862461 |