Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021431.2 | NP_067406.2 | 2 | 1700 | ||
GenBank mRNA | AB041576.1 | - | 2 | 1446 | ||
AK036077.1 | - | 2 | 1031 | |||
AK053310.1 | - | 2 | 1480 | |||
AY152853.1 | - | 2 | 1446 | |||
BC071274.1 | - | 2 | 1504 | 6331160 |