Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253832.1 | NP_001240761.1 | 5 | 703 | ||
NM_021438.4 | NP_067413.1 | 5 | 703 | |||
GenBank mRNA | AF270700.1 | - | 5 | 616 | ||
AK008093.1 | - | 5 | 694 | |||
AK012629.1 | - | 5 | 686 | |||
AK019399.1 | - | 5 | 674 | |||
AK028029.1 | - | 5 | 694 | |||
AK086610.1 | - | 2 | 270 | |||
AK207510.1 | - | 1 | 70 | |||
AK208732.1 | - | 1 | 70 | |||
AK210274.1 | - | 1 | 70 | |||
BC031716.1 | - | 5 | 673 | 3969355 | ||
BC049284.1 | - | 5 | 681 | 6395407 |