Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021439.2 | NP_067414.2 | 3 | 1206 | ||
GenBank mRNA | AB030378.1 | - | 3 | 1358 | ||
AJ289133.1 | - | 3 | 1214 | |||
BC137629.1 | - | 3 | 954 | 8860648 | ||
BC144793.1 | - | 3 | 939 | 9053341 |