Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005568.4 | NP_005559.2 | 2 | 1045 | ||
GenBank mRNA | AK298253.1 | - | 2 | 688 | ||
AK308646.1 | - | 2 | 504 | |||
BC020470.1 | - | 2 | 1045 | |||
BC101674.1 | - | 2 | 414 | 8069180 | ||
BC111936.1 | - | 2 | 414 | 8327404 | ||
U14755.1 | - | 2 | 754 |