Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005568.4 | NP_005559.2 | 3 | 1324 | ||
GenBank mRNA | AK298253.1 | - | 3 | 925 | ||
AK308646.1 | - | 3 | 783 | |||
BC020470.1 | - | 3 | 1324 | |||
BC101674.1 | - | 3 | 693 | 8069180 | ||
BC111936.1 | - | 3 | 693 | 8327404 | ||
U14755.1 | - | 3 | 1033 |