Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021451.2 | NP_067426.1 | 2 | 316 | ||
GenBank mRNA | AB041230.1 | - | 2 | 316 | ||
AK043856.1 | - | 2 | 347 | |||
AK088556.1 | - | 2 | 326 | |||
AK143990.1 | - | 2 | 388 | |||
AK169914.1 | - | 2 | 360 | |||
BC050821.1 | - | 2 | 303 | 6517820 |