Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021453.4 | NP_067428.2 | 1, 2 | 145 | ||
GenBank mRNA | AF240776.1 | - | 1, 2 | 54 | ||
AK004109.1 | - | 1, 2 | 84 | |||
AK053965.1 | - | 1, 2 | 146 | |||
AK168924.1 | - | 1, 2 | 84 | |||
BC119523.2 | - | 1, 2 | 103 | 40056066 |